OriGene
Manufacturer Code:UM800148CF
Catalog # 50-168-5422
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | UMAB258 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-280 of human MSH6 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA mismatch repair protein Msh6; G/T mismatch-binding protein; GTBP; GTBP GTMBP HNPCC5 HSAP p160; hMSH6; MutS protein homolog 6; MutS-alpha 160 kDa subunit; p160; sperm-associated protein
Gene Aliases: GTBP; GTMBP; HNPCC5; HSAP; MSH6; p160
UniProt ID: (Human) P52701
Entrez Gene ID: (Human) 2956
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.