Novus Biologicals
Manufacturer Code:NBP28926550UL
Catalog # NB020015
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 35 |
Immunogen | A synthetic peptide corresponding to the center region of the Human MSH6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA mismatch repair protein Msh6; DNA mismatch repair protein Msh6 G/T mismatch-binding protein GTBPHNPCC5 GTMBP hMSH6 HSAP mutS (E. coli) homolog 6 mutS homolog 6 (E. coli) MutS-alpha 160 kDa subunit p160 sperm-associated protein; G/T mismatch-binding protein; GTBP; hMSH6; MutS protein homolog 6; MutS-alpha 160 kDa subunit; p160; sperm-associated protein
Gene Aliases: GTBP; GTMBP; HNPCC5; HSAP; MSH6; p160
UniProt ID: (Human) P52701
Entrez Gene ID: (Human) 2956
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.