Proteintech
Manufacturer Code:181201AP150UL
Catalog # 50-173-1997
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MSH6 Fusion Protein Ag12645 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA mismatch repair protein Msh6; G/T mismatch binding protein GTBP GTMBP hMSH6 HNPCC5 HSAP MSH6 MutS alpha 160 kDa subunit mutS homolog 6 (E. coli) p160; G/T mismatch-binding protein; GTBP; hMSH6; mutS homolog 6; MutS protein homolog 6; MutS-alpha 160 kDa subunit; p160; rCG61559-like; sperm-associated protein
Gene Aliases: AU044881; AW550279; GTBP; GTMBP; HNPCC5; HSAP; MSH6; p160
UniProt ID: (Human) P52701, (Mouse) P54276
Entrez Gene ID: (Rat) 100360342, (Human) 2956, (Mouse) 17688
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.