Invitrogen
Manufacturer Code:MA535708
Catalog # PIMA535708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1285 |
Immunogen | A synthesized peptide derived from human MSH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AI788990 COCA1 DNA mismatch repair protein MSH2 FCC1 hMSH2 HNPCC HNPCC1 LCFS2 MSH2 Msh2p MutS protein homolog 2 O0935 PMS5 YOL090W; DNA mismatch repair protein Msh2; hMSH2; mismatch repair protein; mutS homolog 2, colon cancer, nonpolyposis type 1; MutS protein homolog 2
Gene Aliases: AI788990; COCA1; FCC1; HNPCC; HNPCC1; LCFS2; MSH2
UniProt ID: (Human) P43246, (Mouse) P43247, (Rat) P54275
Entrez Gene ID: (Human) 4436, (Mouse) 17685, (Rat) 81709
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.