Invitrogen
Manufacturer Code:PA517944
Catalog # PIPA517944
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (PLKSDWNSLDGIIR) corresponding to the C-terminus amino acids of MSC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ABF-1; Activated B-cell factor 1; activated B-cell factor 1, homolog of mouse musculin; activated B-cell factor-1; bHLHa22; Class A basic helix-loop-helix protein 22; MSC musculin (activated B-cell factor-1) ABF1 MYOR ABF-1 musculin activated B-cell factor-1 activated B-cell factor 1 homolog of mouse musculin bHLHa22; Musculin
Gene Aliases: ABF-1; ABF1; BHLHA22; MSC; MYOR
UniProt ID: (Human) O60682
Entrez Gene ID: (Human) 9242
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.