Proteintech
Manufacturer Code:250281AP150UL
Catalog # 50-173-4680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MSC Fusion Protein Ag13489 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABF 1 ABF1 Activated B cell factor 1 bHLHa22 MSC Musculin MYOR; ABF-1; Activated B-cell factor 1; activated B-cell factor 1, homolog of mouse musculin; activated B-cell factor-1; bHLHa22; Class A basic helix-loop-helix protein 22; Musculin; Myogenic repressor
Gene Aliases: ABF-1; ABF1; BHLHA22; MSC; MYOR
UniProt ID: (Human) O60682, (Mouse) O88940
Entrez Gene ID: (Human) 9242, (Mouse) 17681
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.