Invitrogen
Manufacturer Code:PA577558
Catalog # PIPA577558
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)DPKHSSVDRSKLH corresponding to amino acid residues 211-223 of rat MRS2 magnesium transporter |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Magnesium transporter MRS2 homolog, mitochondrial; MRS2 magnesium homeostasis factor homolog; MRS2 Magnesium Transporter; MRS2-like protein; MRS2-like, magnesium homeostasis factor; putative magnesium transporter; RPT protein similar to yeast MRS2
Gene Aliases: Gm902; HPT; MRS2; MRS2L; RPT
UniProt ID: (Human) Q9HD23, (Rat) Q9ET09, (Mouse) Q5NCE8
Entrez Gene ID: (Human) 57380, (Rat) 79032, (Mouse) 380836
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.