OriGene
Manufacturer Code:TA806081
Catalog # 50-168-0961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7F1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 56-262 of human MRRF produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MRFF MTRRF RRF; Ribosome-recycling factor, mitochondrial; Ribosome-releasing factor, mitochondrial; RRF
Gene Aliases: MRFF; MRRF; MTRRF; RRF
UniProt ID: (Human) Q96E11
Entrez Gene ID: (Human) 92399
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.