Invitrogen
Manufacturer Code:PA572397
Catalog # PIPA572397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | region 191-220 aa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MRFF MTRRF RRF ribosome-recycling factor mitochondrial ribosome-releasing factor mitochondrial; Ribosome-recycling factor, mitochondrial; Ribosome-releasing factor, mitochondrial; RRF
Gene Aliases: 2400002D02Rik; MRFF; MRRF; MTRRF; RRF
UniProt ID: (Human) Q96E11, (Mouse) Q9D6S7, (Rat) Q5RKI9
Entrez Gene ID: (Human) 92399, (Mouse) 67871, (Rat) 311903
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.