OriGene
Manufacturer Code:CF505232
Catalog # 50-166-3485
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4C4 |
Immunogen | Full length human recombit protein of human MRPS34 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 28S ribosomal protein S34, mitochondrial; COXPD32 MRP-S12 MRP-S34 MRPS12; mitochondrial 28S ribosomal protein S34; Mitochondrial small ribosomal subunit protein mS34; MRP-S34; S34mt
Gene Aliases: MRP-S12; MRP-S34; MRPS12; MRPS34
UniProt ID: (Human) P82930
Entrez Gene ID: (Human) 65993
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.