Invitrogen
Manufacturer Code:PA524471
Catalog # PIPA524471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 4-32 amino acids from the N-terminal region of human RT34 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28S ribosomal protein S34, mitochondrial; mitochondrial 28S ribosomal protein S34; Mitochondrial small ribosomal subunit protein mS34; MRP-S12 MRP-S34 MRPS12 28S ribosomal protein S34 mitochondrial S34mt; MRP-S34; S34mt; T-complex expressed gene 2 protein
Gene Aliases: 0610007F04Rik; 5330430D13Rik; AV001970; MRP-S12; MRP-S34; MRPS12; MRPS34; Tce2
UniProt ID: (Human) P82930, (Mouse) Q9JIK9
Entrez Gene ID: (Human) 65993, (Mouse) 79044
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.