OriGene
Manufacturer Code:TA800223
Catalog # 50-167-8183
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 151-414 of human MRPS27 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 28S ribosomal protein S27, mitochondrial; mitochondrial 28S ribosomal protein S27; Mitochondrial ribosomal protein S27; Mitochondrial small ribosomal subunit protein mS27; MRP-S27; MRP-S27 S27mt
Gene Aliases: KIAA0264; MRP-S27; MRPS27; S27mt
UniProt ID: (Human) Q92552
Entrez Gene ID: (Human) 23107
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.