Proteintech
Manufacturer Code:163591AP150UL
Catalog # 50-173-0891
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MRPS21 Fusion Protein Ag8343 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28S ribosomal protein S21, mitochondrial; MDS016 MRP S21 MRPS21 RPMS21 S21mt; mitochondrial 28S ribosomal protein S21; Mitochondrial small ribosomal subunit protein bS21m; MRP-S21; S21mt
Gene Aliases: 1810031B19Rik; MDS016; MRP-S21; MRPS21; RPMS21
UniProt ID: (Human) P82921, (Mouse) P58059
Entrez Gene ID: (Human) 54460, (Mouse) 66292, (Rat) 689432
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.