Invitrogen
Manufacturer Code:PA568060
Catalog # PIPA568060
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human MRPS18B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28S ribosomal protein S18-2, mitochondrial; 28S ribosomal protein S18b, mitochondrial; C6orf14 HSPC183 HumanS18a MRP-S18-2 MRPS18-2 PTD017 S18amt 28S ribosomal protein S18-2 mitochondrial 28S ribosomal protein S18b mitochondrial MRP-S18-b S18mt-b mitochondrial ribosomal protein S18-2 mrps18-b; mitochondrial ribosomal protein S18-2; Mitochondrial small ribosomal subunit protein bS18b; Mitochondrial small ribosomal subunit protein mS40; MRP-S18-2; MRP-S18-b; mrps18-b; S18mt-b
Gene Aliases: 2400002C15Rik; C6orf14; HSPC183; HumanS18a; MRP-S18-2; MRPS18-2; MRPS18B; PTD017; S18amt
UniProt ID: (Human) Q9Y676, (Mouse) Q99N84
Entrez Gene ID: (Human) 28973, (Rat) 294230, (Mouse) 66973
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.