Novus Biologicals
Manufacturer Code:NBP215024
Catalog # NBP215024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human MRPS18B. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28S ribosomal protein S18-2 mitochondrial C6orf1428S ribosomal protein S18b mitochondrial HSPC183 HumanS18a mitochondrial ribosomal protein S18-2 mitochondrial ribosomal protein S18B MRP-S18-2 MRPS18-2DKFZp564H0223 Mrps18-b MRP-S18-b PTD017 S18amt S18mt-b; 28S ribosomal protein S18-2, mitochondrial; 28S ribosomal protein S18b, mitochondrial; mitochondrial ribosomal protein S18-2; Mitochondrial small ribosomal subunit protein bS18b; Mitochondrial small ribosomal subunit protein mS40; MRP-S18-2; MRP-S18-b; mrps18-b; S18mt-b
Gene Aliases: C6orf14; HSPC183; HumanS18a; MRP-S18-2; MRPS18-2; MRPS18B; PTD017; S18amt
UniProt ID: (Human) Q9Y676
Entrez Gene ID: (Human) 28973
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.