Invitrogen
Manufacturer Code:PA568059
Catalog # PIPA568059
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human MRPS15. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28S ribosomal protein S15, mitochondrial; Mitochondrial small ribosomal subunit protein uS15m; MRP-S15; RPMS15; S15mt
Gene Aliases: 1500003E24Rik; 2410002B11Rik; DC37; MPR-S15; Mprs15; MRPS15; RPMS15; S15mt
UniProt ID: (Human) P82914, (Mouse) Q9DC71
Entrez Gene ID: (Human) 64960, (Mouse) 66407
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.