Novus Biologicals
Manufacturer Code:NBP287833
Catalog # NB035455
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human MRPS15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28S ribosomal protein S15, mitochondrial; FLJ1156428S ribosomal protein S15 mitochondrial mitochondrial ribosomal protein S15 MPR-S15 MRP-S15 RPMS15 S15mt; Mitochondrial small ribosomal subunit protein uS15m; MRP-S15
Gene Aliases: DC37; MPR-S15; MRPS15; RPMS15; S15mt
UniProt ID: (Human) P82914
Entrez Gene ID: (Human) 64960
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.