Abnova
Manufacturer Code:H00064960K
Catalog # 89-208-788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human MRPS15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human MRPS15 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 28S ribosomal protein S15, mitochondrial; DC37 FLJ11564 MPR-S15 RPMS15 S15mt; Mitochondrial small ribosomal subunit protein uS15m; MRP-S15
Gene Aliases: DC37; MPR-S15; MRPS15; RPMS15; S15mt
UniProt ID: (Human) P82914
Entrez Gene ID: (Human) 64960
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.