Invitrogen
Manufacturer Code:PA590459
Catalog # PIPA590459
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-279 of human MRPL46 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 39S ribosomal protein L46, mitochondrial; L46mt; MRP-L46
Gene Aliases: 3110052F15Rik; C15orf4; C80107; LIECG2; Mrpl46; P2ECSL
UniProt ID: (Mouse) Q9EQI8
Entrez Gene ID: (Mouse) 67308
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.