Novus Biologicals
Manufacturer Code:NBP237393
Catalog # NBP237393
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3H6G11 |
Immunogen | Purified recombinant fragment of human MRPL42 (AA: 142-203) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 28S ribosomal protein S32, mitochondrial; 39S ribosomal protein L31 mitochondrial 39S ribosomal protein L42 mitochondrial HSPC20428S ribosomal protein S32 mitochondrial L31mt mitochondrial ribosomal protein L42 mitochondrial ribosomal protein S32 MRPL31L42mt MRP-L31MRP-S32 MRPS32MRP-L42 PTD007 RPML31S32mt; 39S ribosomal protein L31, mitochondrial; 39S ribosomal protein L42, mitochondrial; L31mt; L42mt; Mitochondrial large ribosomal subunit protein mL42
Gene Aliases: HSPC204; L31MT; L42MT; MRP-L31; MRP-L42; MRP-S32; MRPL31; MRPL42; MRPS32; PTD007; RPML31; S32MT
UniProt ID: (Human) Q9Y6G3
Entrez Gene ID: (Human) 28977
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.