Invitrogen
Manufacturer Code:PA532108
Catalog # PIPA532108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 115 and 288 of Human MRPL39 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 39S ribosomal protein L39, mitochondrial; 39S ribosomal protein L5, mitochondrial; C21orf92 L39mt MRP-L5 MRPL5 PRED22 PRED66 RPML5 39S ribosomal protein L39 mitochondrial 39S ribosomal protein L5 mitochondrial L5mt MRP-L39; L39mt; L5mt; Mitochondrial large ribosomal subunit protein mL39; MRP-L39
Gene Aliases: C21orf92; L39mt; MRP-L5; MRPL39; MRPL5; MSTP003; PRED22; PRED66; RPML5
UniProt ID: (Human) Q9NYK5
Entrez Gene ID: (Human) 54148
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.