Novus Biologicals
Manufacturer Code:NBP186815
Catalog # NBP186815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ERIVKLHRIGDFIDVSEGPLIPRTSICFQYEVSAVHNLQPTQPSLIRRFQGVSLPVHLRAHFTIWDKLLERSRKMVTEDQSKAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 39S ribosomal protein L39, mitochondrial; 39S ribosomal protein L5, mitochondrial; C21orf92PRED66 FLJ2045139S ribosomal protein L39 mitochondrial L39mt39S ribosomal protein L5 mitochondrial MGC104174 MGC3400 mitochondrial ribosomal protein L39 MRP-L5L5mt MRPL5MSTP003 PRED22 RPML5MRP-L39; L39mt; L5mt; Mitochondrial large ribosomal subunit protein mL39; MRP-L39
Gene Aliases: C21orf92; L39mt; MRP-L5; MRPL39; MRPL5; MSTP003; PRED22; PRED66; RPML5
UniProt ID: (Human) Q9NYK5
Entrez Gene ID: (Human) 54148
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.