Proteintech
Manufacturer Code:281651AP150UL
Catalog # 50-173-5825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MRPL39 Fusion Protein Ag27154 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 39S ribosomal protein L39, mitochondrial; 39S ribosomal protein L5, mitochondrial; C21orf92 FLJ20451 L39mt L5mt MRP L39 MRP L5 MRPL39 MRPL5 MSTP003 PRED22 PRED66 RPML5; L39mt; L5mt; Mitochondrial large ribosomal subunit protein mL39; mitochondrial ribosomal protein L39; MRP-L39; ribosomal protein, mitochondrial, L5
Gene Aliases: C21orf8; C21orf92; L39mt; MRP-L5; MRPL39; MRPL5; MSTP003; ORF22; PRED22; PRED66; RPML5
UniProt ID: (Human) Q9NYK5, (Mouse) Q9JKF7
Entrez Gene ID: (Human) 54148, (Rat) 684304, (Mouse) 27393
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.