Invitrogen
Manufacturer Code:PA5103523
Catalog # PIPA5103523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MRPL28 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110015G04Rik 39S ribosomal protein L28 mitochondrial HGNC6756 L28mt MAAT 1 MAAT1 Melanoma antigen p15 melanoma-associated antigen recognised by cytotoxic T lymphocytes melanoma-associated antigen recognized by T lymphocytes melanoma-associated antigen recognized by T-lymphocytes MGC8499 Mitochondrial large ribosomal subunit protein bL28m mitochondrial ribosomal protein L28 MRP L28 MRPL 28 MRPL28 MRP-L28 p15; 39S ribosomal protein L28, mitochondrial; L28mt; Melanoma antigen p15; melanoma-associated antigen recognised by cytotoxic T lymphocytes; melanoma-associated antigen recognized by T lymphocytes; Melanoma-associated antigen recognized by T-lymphocytes; Mitochondrial large ribosomal subunit protein bL28m; MRP-L28
Gene Aliases: 1110015G04Rik; L28mt; MAAT1; MRP-L28; MRPL28; p15
UniProt ID: (Human) Q13084, (Mouse) Q9D1B9
Entrez Gene ID: (Human) 10573, (Mouse) 68611
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.