Proteintech
Manufacturer Code:155431AP150UL
Catalog # 50-173-0301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MRPL11 Fusion Protein Ag7899 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 39S ribosomal protein L11, mitochondrial; CGI 113 L11MT MRP L11 MRPL11; L11mt; Mitochondrial large ribosomal subunit protein uL11m; MRP-L11
Gene Aliases: 2410001P07Rik; CGI-113; L11MT; MRP-L11; MRPL11
UniProt ID: (Human) Q9Y3B7, (Mouse) Q9CQF0, (Rat) Q5XIE3
Entrez Gene ID: (Human) 65003, (Mouse) 66419, (Rat) 293666
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.