Novus Biologicals
Manufacturer Code:NBP272779C
Catalog # NBP272779C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B4 |
Immunogen | Full length recombinant protein of human MRPL10 (NP_660298) produced in E.coli. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 39S ribosomal protein L10, mitochondrial; 39S ribosomal protein L8, mitochondrial; L10mt; L10mt MGC17973 mitochondrial ribosomal protein L10 MRP-L10L8mt MRP-L839S ribosomal protein L10 mitochondrial MRPL839S ribosomal protein L8 mitochondrial RPML8L10MT; L8mt; Mitochondrial large ribosomal subunit protein uL10m
Gene Aliases: L10MT; MRP-L10; MRP-L8; MRPL10; MRPL8; RPML8
UniProt ID: (Human) Q7Z7H8
Entrez Gene ID: (Human) 124995
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.