Invitrogen
Manufacturer Code:PA5111033
Catalog # PIPA5111033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human MRP8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60B8Ag AI323541 B8Ag Caga Calgranulin-A Calprotectin L1L subunit CFAG CGLA Chemotactic cytokine CP-10 CP-10 Cystic fibrosis antigen L1Ag Leukocyte L1 complex light chain MA387 MIF Migration inhibitory factor-related protein 8 MRP8 MRP-8 NIF P8 Pro-inflammatory S100 cytokine Protein S100-A8 S100 A8 S100 calcium-binding protein A8 S100a8 Urinary stone protein band A; calgranulin A; Calgranulin-A; Calprotectin L1L subunit; CFAG; Cystic fibrosis antigen; Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8; MRP-8; Protein S100-A8; S100 calcium-binding protein A8; S100 calcium-binding protein A8 (calgranulin A); Urinary stone protein band A
Gene Aliases: 60B8AG; CAGA; CFAG; CGLA; CP-10; L1Ag; MA387; MIF; MRP8; NIF; P8; S100A8
UniProt ID: (Human) P05109
Entrez Gene ID: (Human) 6279
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.