Invitrogen
Manufacturer Code:MA126542
Catalog # PIMA126542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | M6II-31 |
Immunogen | Bacterial fusion protein of human MRP 6 containing amino acids 764-964 spanning the putative 12th transmembrane region as well as predicted internal and external regions of the protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ABC34 ARA EST349056 MLP1 MOATE MRP6 PXE PXE1 URG7; Anthracycline resistance-associated protein; ATP-binding cassette sub-family C member 6; ATP-binding cassette, sub-family C (CFTR/MRP), member 6; MOAT-E; Multi-specific organic anion transporter E; Multidrug resistance-associated protein 6
Gene Aliases: ABC34; ABCC6; ARA; EST349056; GACI2; MLP1; MOAT-E; MOATE; MRP6; PXE; PXE1; URG7
UniProt ID: (Human) O95255
Entrez Gene ID: (Human) 368
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.