Invitrogen
Manufacturer Code:PA576689
Catalog # PIPA576689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human MRP5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC 33ABC33ABCC 5ABCC5ATP binding cassette sub family C (CFTR/MRP) member 5ATP binding cassette sub family C member 5ATP-binding cassette sub-family C member 5Canalicular multispecific organic anion transporter CDKFZp686C1782EST277145MOAT CMOAT-CMOATCMRP 5MRP5&lowbarHUMANMulti specific organic anion tranporter CMulti-specific organic anion transporter CMultidrug resistance associated protein 5Multidrug resistance-associated protein 5pABC 11pABC11SMRP; ATP-binding cassette sub-family C (CFTR/MRP) member 5a; ATP-binding cassette sub-family C member 5; ATP-binding cassette, sub-family C (CFTR/MRP), member 5; ATP-binding cassette, sub-family C (CFTR/MRP), member 5a; ATP-binding cassette, sub-family C (CFTR/MRP), member 5b; ATP-binding cassette, sub-family C, member 5; canalicular multispecific organic anion transporter C; MOAT-C; Multi-specific organic anion transporter C; Multidrug resistance-associated protein 5; pABC11; SMRP
Gene Aliases: 2900011L11Rik; ABC33; ABCC5; Abcc5a; Abcc5b; AI132311; EST277145; MOAT-C; MOATC; MRP5; pABC11; SMRP
UniProt ID: (Human) O15440, (Rat) Q9QYM0, (Mouse) Q9R1X5
Entrez Gene ID: (Human) 10057, (Rat) 116721, (Mouse) 27416
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.