Invitrogen
Manufacturer Code:PA5102678
Catalog # PIPA5102678
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ABCC5 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900011L11Rik ABC 33 ABC33 ABCC 5 ABCC5 Abcc5a Abcc5b AI132311 ATP binding cassette subfamily C member 5 ATP-binding cassette sub-family C (CFTR/MRP) member 5a ATP-binding cassette sub-family C member 5 ATP-binding cassette subfamily C (CFTR/MRP) member 5 ATP-binding cassette sub-family C (CFTR/MRP) member 5 ATP-binding cassette sub-family C (CFTR/MRP) member 5a ATP-binding cassette sub-family C (CFTR/MRP) member 5b ATP-binding cassette sub-family C member 5 canalicular multispecific organic anion transporter C DKFZp686C1782 EST277145 MOAT C MOATC MOAT-C MRP 5 MRP5 Multidrug resistance-associated protein 5 Multi-specific organic anion transporter C pABC 11 pABC11 SMRP; ATP-binding cassette sub-family C (CFTR/MRP) member 5a; ATP-binding cassette sub-family C member 5; ATP-binding cassette, sub-family C (CFTR/MRP), member 5; ATP-binding cassette, sub-family C (CFTR/MRP), member 5a; ATP-binding cassette, sub-family C (CFTR/MRP), member 5b; ATP-binding cassette, sub-family C, member 5; canalicular multispecific organic anion transporter C; MOAT-C; Multi-specific organic anion transporter C; Multidrug resistance-associated protein 5; pABC11; SMRP
Gene Aliases: 2900011L11Rik; ABC33; ABCC5; Abcc5a; Abcc5b; AI132311; EST277145; MOAT-C; MOATC; MRP5; pABC11; SMRP
UniProt ID: (Human) O15440, (Rat) Q9QYM0, (Mouse) Q9R1X5
Entrez Gene ID: (Human) 10057, (Rat) 116721, (Mouse) 27416
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.