Invitrogen
Manufacturer Code:PA518315
Catalog # PIPA518315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (CNGQPSTLTIFETAL) corresponding to the C-terminus amino acids of ABCC4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC-tranporter; ABCC4 MRP4 MOATB MOAT-B EST170205 ATP-binding cassette sub-family C (CFTR/MRP) member 4 canalicular multispecific organic anion transporter (ABC superfamily) Multidrug Resistance Protein 4 RP11-74A12.1 ATP-binding cassette sub-family C member 4 bA464I2.1 (ATP-binding cassette sub-family C (CFTR/MRP) member 4) canalicular multispecific organic anion transporter (ABC superfamily) multidrug resistance-associated protein 4 multispecific organic anion transporter B; ATP-binding cassette protein C4; ATP-binding cassette sub-family C member 4; ATP-binding cassette subfamily C member 4; ATP-binding cassette, sub-family C (CFTR/MRP), member 4; bA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4); canalicular multispecific organic anion transporter (ABC superfamily); MOAT-B; MRP/cMOAT-related ABC transporter; Multi-specific organic anion transporter B; Multidrug resistance-associated protein 4
Gene Aliases: ABCC4; MOAT-B; MOATB; MRP4
UniProt ID: (Human) O15439, (Rat) F1M3J4
Entrez Gene ID: (Human) 10257, (Rat) 170924
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.