Novus Biologicals
Manufacturer Code:NBP237923
Catalog # NBP237923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: FLPQTDFIIVLADGQVSEMGPYPALLQRNGSFANFLCNYAPDEDQGHLEDSWTALEGAEDKEALLIEDTLSNHTDLTDNDPVTYVVQKQFMRQLSALSSDGEGQGRPVPRRHLGPSEKVQVTEAKADGALT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC31 ATP-binding cassette sub-family C member 3 ATP-binding cassette sub-family C (CFTR/MRP) member 3 canalicular multispecific organic anion transporter 2 canicular multispecific organic anion transporter cMOAT2 EC 3.6.3 EC 3.6.3.44 EC 6.3.2.4 MLP2DKFZp686E22157 MRP3MOAT-DEST90757 multidrug resistance associated protein Multidrug resistance-associated protein 3 Multi-specific organic anion transporter D; ATP-binding cassette sub-family C member 3; ATP-binding cassette, sub-family C (CFTR/MRP), member 3; Canalicular multispecific organic anion transporter 2; canicular multispecific organic anion transporter; MOAT-D; Multi-specific organic anion transporter D; multidrug resistance associated protein; Multidrug resistance-associated protein 3
Gene Aliases: ABC31; ABCC3; CMOAT2; EST90757; MLP2; MOAT-D; MRP3
UniProt ID: (Human) O15438
Entrez Gene ID: (Human) 8714
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.