Invitrogen
Manufacturer Code:MA135603
Catalog # 01-671-041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M2I-4 |
Immunogen | Proprietary immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ABCC2 cMOAT canalicular multi-organic anion transporter ABC30; ATP-binding cassette sub-family C member 2; ATP-binding cassette, sub-family C (CFTR/MRP), member 2; Canalicular multidrug resistance protein; Canalicular multispecific organic anion transporter 1; Multidrug resistance-associated protein 2
Gene Aliases: ABC30; ABCC2; CMOAT; CMOAT1; CMRP; DJS; MRP2
UniProt ID: (Human) Q92887
Entrez Gene ID: (Human) 1244
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.