Invitrogen
Manufacturer Code:PA575534
Catalog # PIPA575534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 1036-1081 of Human MRP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-binding cassette sub-family C member 2; ATP-binding cassette, sub-family C (CFTR/MRP), member 2; calicular multispecific organic anion transporter; Canalicular multidrug resistance protein; Canalicular multispecific organic anion transporter 1; Canalicular multispecific organic anion transporter 1 ATP-binding cassette sub-family C member 2 Canalicular multidrug resistance protein Multidrug resistance-associated protein 2 ABCC2 CMOAT CMOAT1 CMRP; multidrug resistance associated protein 2; multidrug resistance protein 2; Multidrug resistance-associated protein 2
Gene Aliases: ABC30; ABCC2; AI173996; CMOAT; CMOAT1; CMRP; DJS; MRP2
UniProt ID: (Human) Q92887, (Rat) Q63120, (Mouse) Q8VI47
Entrez Gene ID: (Human) 1244, (Rat) 25303, (Mouse) 12780
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.