Invitrogen
Manufacturer Code:PA523652
Catalog # PIPA523652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 767-793 amino acids from the central region of human ABCC10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-binding cassette sub-family C member 10; ATP-binding cassette, sub-family C (CFTR/MRP), member 10; EST182763 MRP7 SIMRP7 ATP-binding cassette sub-family C member 10 multidrug resistance-associated protein 7; Multidrug resistance-associated protein 7
Gene Aliases: ABCC10; EST182763; MRP7; SIMRP7
UniProt ID: (Human) Q5T3U5
Entrez Gene ID: (Human) 89845
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.