Novus Biologicals
Manufacturer Code:NB400156IR
Catalog # NB400156IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | IU2H10 |
Immunogen | Recombinant peptide (a.a. 1-33) of human MRP1. [UniProt# P33527]. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ABC29 ABCC ATP-binding cassette sub-family C member 1 ATP-binding cassette sub-family C (CFTR/MRP) member 1 EC 3.6.3 EC 3.6.3.44 GS-X Leukotriene C(4) transporter LTC4 transporter MRP1DKFZp781G125 MRPDKFZp686N04233 multidrug resistance associated protein 1 multidrug resistance protein multidrug resistance-associated protein 1 multiple drug resistance protein 1 multiple drug resistance-associated protein; ATP-binding cassette sub-family C member 1; ATP-binding cassette transporter variant ABCC1delta-ex13; ATP-binding cassette transporter variant ABCC1delta-ex13&14; ATP-binding cassette transporter variant ABCC1delta-ex25; ATP-binding cassette transporter variant ABCC1delta-ex25&26; ATP-binding cassette, sub-family C (CFTR/MRP), member 1; Glutathione-S-conjugate-translocating ATPase ABCC1; Leukotriene C(4) transporter; LTC4 transporter; Multidrug resistance-associated protein 1
Gene Aliases: ABC29; ABCC; ABCC1; GS-X; MRP; MRP1
UniProt ID: (Human) P33527
Entrez Gene ID: (Human) 4363
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.