Invitrogen
Manufacturer Code:PA541911
Catalog # PIPA541911
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human MRM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 16S rRNA (guanosine(1145)-2'-O)-methyltransferase; 16S rRNA [Gm1145] 2'-O-methyltransferase; CTB-75G16.3; mitochondrial large ribosomal RNA ribose methylase; mitochondrial rRNA methyltransferase 1 homolog; Mitochondrial rRNA methyltransferase 1 homolog (S. cerevisiae); rRNA methyltransferase 1, mitochondrial
Gene Aliases: A530065E19Rik; ENSMUSG00000054212; MRM1; RGD1566232
UniProt ID: (Human) Q6IN84, (Mouse) Q99J25
Entrez Gene ID: (Horse) 100071427, (Dog) 610311, (Rabbit) 100350035, (Bovine) 526282, (Human) 79922, (Guinea pig) 100732290, (Rat) 363661, (Mouse) 217038
Molecular Function:
RNA binding protein
RNA methyltransferase
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.