Novus Biologicals
Manufacturer Code:NBP183597
Catalog # NBP183597
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SCQLLLTILPRRQLPPGLESLNVSVAAGILLHSICSQRKGFPTEGERRQLLQDPQEPSARSEGLSMAQHPGLSS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 16S rRNA (guanosine(1145)-2'-O)-methyltransferase; 16S rRNA [Gm1145] 2'-O-methyltransferase; CTB-75G16.3; EC 2.1.1.- FLJ22578 Mitochondrial large ribosomal RNA ribose methylase mitochondrial rRNA methyltransferase 1 homolog (S. cerevisiae) rRNA methyltransferase 1 mitochondrial; mitochondrial large ribosomal RNA ribose methylase; mitochondrial rRNA methyltransferase 1 homolog; rRNA methyltransferase 1, mitochondrial
Gene Aliases: MRM1
UniProt ID: (Human) Q6IN84
Entrez Gene ID: (Human) 79922
Molecular Function: RNA binding protein RNA methyltransferase methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.