Invitrogen
Manufacturer Code:PA568858
Catalog # PIPA568858
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse MRI1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: M1Pi; M1Pi {ECO:0000255|HAMAP-Rule:MF_03119}; Mediator of RhoA-dependent invasion; Methylthioribose-1-phosphate isomerase; methylthioribose-1-phosphate isomerase homolog; Methylthioribose-1-phosphate isomerase homolog (S. cerevisiae) MGC3207 MTNA Ypr118w; methylthioribose-1-phosphate isomerase {ECO:0000255|HAMAP-Rule:MF_03119}; MRI1; MTR-1-P isomerase; MTR-1-P isomerase {ECO:0000255|HAMAP-Rule:MF_03119}; S-methyl-5-thioribose-1-phosphate isomerase; S-methyl-5-thioribose-1-phosphate isomerase 1; S-methyl-5-thioribose-1-phosphate isomerase {ECO:0000255|HAMAP-Rule:MF_03119}; Translation initiation factor eIF-2B subunit alpha/beta/delta-like protein; translation initiation factor eIF-2B subunit alpha/beta/delta-like protein {ECO:0000255|HAMAP-Rule:MF_03119}
Gene Aliases: 2410018C20Rik; CH73-131E21.1; M1Pi; MRDI; MRI1; MTNA; RGD1307789; UNQ6390/PRO21135; Ypr118w; zgc:172216
UniProt ID: (Bovine) Q2NL31, (Human) Q9BV20, (Rat) Q5HZE4, (Mouse) Q9CQT1, (Zebrafish) A9JRE2
Entrez Gene ID: (Bovine) 534734, (Human) 84245, (Dog) 484914, (Horse) 100146396, (Rabbit) 100342612, (Pig) 100525858, (Rat) 288912, (Mouse) 67873, (Zebrafish) 100002302
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.