Abnova
Manufacturer Code:PAB10277
Catalog # 89-114-748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 578-590 of <i>Saccharomyces cerevisiae</i> MRE11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of MRE11.
For Research Use Only
Protein Aliases: Double-strand break repair protein MRE11; MRX complex nuclease subunit; NGS1; RAD58; XRS4
Gene Aliases: MRE11; NGS1; RAD58; XRS4; YM9959.06C; YMR224C
UniProt ID: (Baker's yeast) P32829
Entrez Gene ID: (Baker's yeast) 855264
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.