Proteintech
Manufacturer Code:107441AP150UL
Catalog # 50-172-6559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MRE11 Fusion Protein Ag1105 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AT-like disease; ATLD HNGS1 MRE11 MRE11 homolog 1 MRE11 homolog A MRE11A MRE11B; DNA recombination and repair protein; Double-strand break repair protein MRE11; Double-strand break repair protein MRE11A; endo/exonuclease Mre11; Meiotic recombination 11 homolog 1; Meiotic recombination 11 homolog A; MmMRE11A; MRE11 double strand break repair nuclease A; MRE11 homolog 1; MRE11 homolog A; MRE11 homolog, double strand break repair nuclease A; MRE11 meiotic recombination 11 homolog A; MRE11 meiotic recombination 11-like protein A
Gene Aliases: ATLD; HNGS1; MRE11; MRE11A; MRE11B
UniProt ID: (Human) P49959, (Rat) Q9JIM0, (Mouse) Q61216
Entrez Gene ID: (Human) 4361, (Rat) 64046, (Mouse) 17535
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.