OriGene
Manufacturer Code:TA811858
Catalog # 50-168-4296
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9G4 |
Immunogen | Full length human recombit protein of human MRC2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-type lectin domain family 13 member E; C-type mannose receptor 2; CD280; CD280 CLEC13E ENDO180 UPARAP; endocytic receptor (macrophage mannose receptor family); Endocytic receptor 180; Macrophage mannose receptor 2; mannose receptor, C type 2; UPAR-associated protein; Urokinase-type plasminogen activator receptor-associated protein
Gene Aliases: CD280; CLEC13E; ENDO180; KIAA0709; MRC2; UPARAP
UniProt ID: (Human) Q9UBG0
Entrez Gene ID: (Human) 9902
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.