Proteintech
Manufacturer Code:142131AP150UL
Catalog # 50-172-9287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MRAS Fusion Protein Ag5433 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: M RAs MRAS muscle RAS oncogene homolog R RAS3 Ras related protein M Ras Ras related protein R Ras3 RRAS3; Muscle and microspikes Ras; Ras-related protein M-Ras; Ras-related protein R-Ras3; X-Ras
Gene Aliases: 2900078C09Rik; AI326250; M-RAs; MRAS; R-RAS3; RRAS3; Xras
UniProt ID: (Human) O14807, (Mouse) O08989
Entrez Gene ID: (Human) 22808, (Mouse) 17532
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.