Invitrogen
Manufacturer Code:PA571793
Catalog # PIPA571793
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 58-86 amino acids from the Central region of human MRAP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MC2R accessory protein 2; Melanocortin-2 receptor accessory protein 2; Melanocortin-2 receptor accessory protein 2 MC2R accessory protein 2 MRAP2 C6orf117
Gene Aliases: bA51G5.2; BMIQ18; C6orf117; MRAP2
UniProt ID: (Human) Q96G30
Entrez Gene ID: (Human) 112609
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.