Invitrogen
Manufacturer Code:PA5113283
Catalog # PIPA5113283
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Melanocortin-2 receptor accessory protein 2 protein (66-205AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA51G5.2 BB633055 BMIQ18 C6orf117 MC2R accessory protein 2 melanocortin 2 receptor accessory protein 2 Melanocortin-2 receptor accessory protein 2 MRAP2 RGD1309873; MC2R accessory protein 2; Melanocortin-2 receptor accessory protein 2
Gene Aliases: bA51G5.2; BB633055; BMIQ18; C6orf117; MRAP2
UniProt ID: (Human) Q96G30, (Mouse) D3Z1Q2
Entrez Gene ID: (Human) 112609, (Mouse) 244958
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.