Invitrogen
Manufacturer Code:PA5106939
Catalog # PIPA5106939
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MRAP corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110025G12Rik B27 C21orf61 FALP Fat cell-specific low molecular weight protein fat tissue-specific low MW protein FGD2 GCCD2 melanocortin 2 receptor accessory protein melanocortin-2 receptor accessory protein Mrap ORF61; B27; Fat cell-specific low molecular weight protein; Fat tissue-specific low MW protein; Melanocortin-2 receptor accessory protein
Gene Aliases: 1110025G12Rik; B27; C21orf61; FALP; FGD2; GCCD2; MRAP; ORF61
UniProt ID: (Human) Q8TCY5, (Mouse) Q9D159
Entrez Gene ID: (Human) 56246, (Mouse) 77037
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.