Proteintech
Manufacturer Code:105721AP150UL
Catalog # 50-172-6416
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MPZP0 Fusion Protein Ag0848 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Charcot-Marie-Tooth neuropathy 1B; CHM CMT1 CMT1B CMT2I CMT2J CMT4E CMTDI3 DSS HMSNIB MPP MPZ Myelin peripheral protein Myelin protein P0 myelin protein zero P0; MPP; Myelin peripheral protein; Myelin protein P0; Myelin protein zero; myelin protein zero (Charcot-Marie-Tooth neuropathy 1B)
Gene Aliases: CHM; CMT1; CMT1B; CMT2I; CMT2J; CMT4E; CMTDI3; CMTDID; DSS; HMSNIB; MPP; MPZ; P-zero; P0
UniProt ID: (Human) P25189, (Rat) P06907, (Mouse) P27573
Entrez Gene ID: (Human) 4359, (Rat) 24564, (Mouse) 17528
Molecular Function:
cell adhesion molecule
ion channel
sodium channel
transporter
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.