Invitrogen
Manufacturer Code:PA598291
Catalog # PIPA598291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Myelin protein P0 protein (30-156AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Charcot-Marie-Tooth neuropathy 1B; MPP; Myelin peripheral protein; Myelin protein P0; Myelin protein zero
Gene Aliases: CHM; CMT1; CMT1B; CMT2I; CMT2J; CMT4E; CMTDI3; CMTDID; DSS; HMSNIB; MPP; MPZ; P0
UniProt ID: (Human) P25189
Entrez Gene ID: (Human) 4359
Molecular Function:
cell adhesion molecule
ion channel
sodium channel
transporter
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.