Invitrogen
Manufacturer Code:PA518773
Catalog # PIPA518773
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DHSRSTKAVSEK) corresponding to the internal amino acids of MPZ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: MPP; Myelin peripheral protein; Myelin protein P0; Myelin protein zero; myelin protein zero (Charcot-Marie-Tooth neuropathy 1B); myelin protein zero myelin protein zero (Charcot-Marie-Tooth neuropathy 1B) MPZ CHM CMT1 CMT1B CMT2I CMT2J CMT4E CMTDI3 DSS HMSNIB MPP P0
Gene Aliases: MPP; Mpz; P0
UniProt ID: (Rat) P06907
Entrez Gene ID: (Rat) 24564
Molecular Function: cell adhesion molecule ion channel sodium channel transporter voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.