Abnova
Manufacturer Code:H00004359A01
Catalog # 89-003-023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | MPZ (AAH06491.1 1 a.a. ~ 258 a.a) full-length recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant MPZ.
For Research Use Only
Protein Aliases: Charcot-Marie-Tooth neuropathy 1B; CHM; CMT1; CMT1B; CMT2I; CMT2J; CMT4E; CMTDI3; DSS; HMSNIB; MPP; Myelin peripheral protein; Myelin protein P0; Myelin protein zero; P0
Gene Aliases: CHM; CMT1; CMT1B; CMT2I; CMT2J; CMT4E; CMTDI3; CMTDID; DSS; HMSNIB; MPP; MPZ; P0
UniProt ID: (Human) P25189
Entrez Gene ID: (Human) 4359
Molecular Function:
cell adhesion molecule
ion channel
sodium channel
transporter
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.